Cryogenic storage vessel, ARPEGE CryoMemo, Air Liquide
Supplier: Air Liquide
ARPEGE55NL103 ARPEGE70NG102 ARPEGE70NG103 ARPEGE70NL1 ARPEGE70NL101 ARPEGE70NL102 ARPEGE70NL103 ARPEGE75NL1 ARPEGE75NL101 ARPEGE75NL103 ARPEGE55NL1 ARPEGE75NL102 ARPEGE170NG103 ARPEGE55NL102 ARPEGE170NG102 ARPEGE55NL101 ARPEGE170NL1 ARPEGE170NL101 ARPEGE170NL102 ARPEGE170NL103 ARPEGE40NL1 ARPEGE40NL101 ARPEGE140NL103 ARPEGE110NG103 ARPEGE110NG102 ARPEGE110NL1 ARPEGE110NL101 ARPEGE110NL102 ARPEGE110NL103 ARPEGE140NG102 ARPEGE140NG103 ARPEGE140NL1 ARPEGE140NL101 ARPEGE140NL102
479-1786EA 10810 EUR
479-1786 478-0752 478-0753 479-1787 478-0754 478-0755 478-0756 479-1788 479-1789 479-1791 479-1783 479-1790 478-0747 479-1785 478-0746 479-1784 479-1781 478-0748 478-0749 478-0750 479-1782 478-0751 478-0745 478-0737 478-0736 479-1779 478-0738 478-0739 478-0740 478-0741 478-0742 479-1780 478-0743 478-0744
Cryogenic storage vessel, ARPEGE CryoMemo, Air Liquide
Dewars and Cryogenic Vessels
These vessels are designed for long term storage of biological samples in straws, cryo-vials and bags. A variety of electronic options on all models rack or canister storage systems for the samples.
Made of aluminum and composite materials.
  • Light weight and long holding time
  • Optimum safety: system designed to secure and keep each storage box in its place in the rack
  • Level display in % and temperature in °C or °F associated with a simple access to all parameters of the containers (level threshold high and low, alarm values...)
  • Visual, audible and graphic
More Product Information