Cold traps, Type KF
Supplier: KGW
1731 1740 17402 1735 17377 1741 17372 17371
478-4422EA 602 EUR
478-4422 KGWA1740 KGWA17402 KGWA1735 KGWA17377 KGWA1741 KGWA17372 KGWA17371
Cold traps, Type KF
Vacuum Traps
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents. Model KF cold traps have a GL 32 glass thread together with a screw-on cap (spout), through which the condensate can easily be poured out of the cold trap. Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
  • Easy to clean
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask made of blue coated metal or aluminium
Order Now
SPECIFICATIONS
Environmentally Preferable