Racks for chest freezers, Comfort
Supplier: TENAK
TE24518 TE24516 TE24514 TE24512 TE24510 TE24508 TE24506 TE24504 TE24500 TE24534 TE24502 TE24566 TE24530 TE24532 TE24564 TE24562 TE24560 TE24544 TE24542 TE24540 TE24538 TE24536 TE24490 TE24491 TE24492 TE24493 TE24494 TE24466 TE24468 TE24470 TE24465 TE24462 TE24464 TE24458 TE24460 TE24495 TE24496 TE24520 TE24522 TE24500B TE24502B TE24504B TE24506B TE24508B TE24510B TE24512B TE24514B TE24516B TE24518B TE24530B TE24532B TE24534B TE24536B TE24538B TE24540B TE24542B TE24544B TE24560B TE24562B TE24564B TE24566B TE24576 TE24570 TE24572 TE24574
479-0794EA 132 EUR
479-0794 479-0793 479-0792 479-0791 479-0790 479-0789 479-0788 479-0787 479-0785 479-0797 479-0786 479-0840 479-0795 479-0796 479-0839 479-0838 479-0837 479-0836 479-0835 479-0834 479-0799 479-0798 479-0861 479-0862 479-0863 479-0864 479-0865 479-1684 479-1685 479-1686 479-1683 479-1681 479-1682 479-1378 479-1680 479-1183 479-1184 479-1185 479-1186 479-1687 479-1688 479-1689 479-1690 479-1691 479-1692 479-1693 479-1694 479-1695 479-1696 479-1697 479-1698 479-1699 479-1700 479-1701 479-1702 479-1703 479-1704 479-1705 479-1706 479-1707 479-1708 479-1712 479-1709 479-1710 479-1711
Racks for chest freezers, Comfort
Racks Cryogenic Box Racks
Stainless steel.
  • Can be used with all brands of freezers
  • Suitable for all standard cryoboxes
  • Racks have spring locks mounted on each shelf to secure the cryoboxes
  • Suitable for cardboard, polypropylene and polycarbonate cryoboxes
The racks are an economical storage system for chest freezers.
Half box racks 50 mm height. The racks can be used with boxes with a max. footprint of 83×83 mm, typically with the 5×5 cryobox.