Cryogenic ampoules, Cryule®, WHEATON®
Supplier: DWK Life Sciences
651486 651463 651483 W651469
479-1127EA 199 EUR
479-1127 479-1119 479-1126 479-2091
Cryogenic ampoules, Cryule®, WHEATON®
Ampoules
All glass ampoules that can be hermetically sealed to preserve sample purity and extend sample shelf life. Ideal for standards packaging, environmental and diagnostic standards, lyophilisation and cryogenic storage.
  • Tamper-evident seal
  • For biological sample preservation in glass
  • Can be used in the vapour phase of liquid nitrogen
  • Special design allows for storage at low temperatures as well as rapid thawing
Made from low extractable borosilicate glass, conform to USP Type I and ASTM E438 Type I, Class A requirements.
A gold band on the stem of theMore Product Information