Geraniol, Supelco®

Supplier: Merck
Warning

Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-ol

MQ100

48798-1ML 48798-5ML
48798-1MLEA 187 EUR
48798-1ML 48798-5ML
Geraniol, Supelco®
Geraniol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00002917
CAS Number: 106-24-1
EINECS: 203-377-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5db19c236d64f8f657ad453ec88e7537c504df8b&vid=147f4df4fdec28d35a4f08f8ddc3e5dcde975aa8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR