Nonidet® P 40 Substitute (NP-40) 10% aqueous solution, Millipore®

Supplier: Merck Millipore Calbiochem

492018-50 492018-500
492018-50EA 282 EUR
492018-50 492018-500
Nonidet® P 40 Substitute (NP-40) 10% aqueous solution, Millipore®
Nonidet® P 40 Substitute (NP-40)
MDL Number: MFCD00132851
CAS Number: 9016-45-9
EINECS: 500-024-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR