Organic reference standard, Neotame, analytical standard, Supelco®
Supplier: Merck
49777-100MG
49777-100MGEA 85.5 EUR
49777-100MG
Organic reference standard, Neotame, analytical standard, Supelco®
Standards Organic Standards
Neotame is a white colored, dipeptide methyl ester, which is found to be a highly potent and non-nutritive sweetener or flavour enhancer and hence finds applications in variety of foods. It is manufactured from aspartame and 3,3-dimethylbutyraldehyde in a one-step chemical synthesis process.
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d2472d9f3feffe871683cbe10bd05bf2bf9a829&vid=3d2472d9f3feffe871683cbe10bd05bf2bf9a829&prid=web&clid=SAPDAM