Brix sugar standards for refractometers, Certipur®, Supelco®

Supplier: Merck

5.00420.0001 5.00430.0001 5.00400.0001 5.00405.0001 5.00410.0001 5.00440.0001 5.00460.0001 5.00450.0001
5.00420.0001EA 206 EUR
5.00420.0001 5.00430.0001 5.00400.0001 5.00405.0001 5.00410.0001 5.00440.0001 5.00460.0001 5.00450.0001
Brix sugar standards for refractometers, Certipur®, Supelco®
Standards Refractive Index Standards

Refractive index standards traceable to SRM from NIST and PTB.


  • Each ampoule contains about 8 ml solution
  • Minimum shelf life of Brix standard 0° is 36 months
  • All other standards minimum shelf life is 24 month
  • Tested for homogeneity and stability
  • Storage at room temperature

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR