DL-α-Tocopherol (Vitamin E), dried powder, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS

5.00859.2500 5.00859.9029
5.00859.2500EA 1070 EUR
5.00859.2500 5.00859.9029
DL-α-Tocopherol (Vitamin E), dried powder, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
DL-α-Tocopherol

Vitamin E (50 g all-rac-α-tocopheryl acetate in 100 g substance)

Formula: C₂₉H₅₀O₂
MDL Number: MFCD00072051
CAS Number: 10191-41-0
EINECS: 233-466-0

Order Now

Specification Test Results

all-rac-α-Tocopheryl acetate content (GC, Ph. Eur.) 50.0 - 57.5 %
all-rac-α-Tocopheryl acetate content (GC, USP) 49.0 - 57.5 %
Identity (TLC) conforms
Identity (GC) conforms
Optical rotation α 20/D (all-rac-α-Tocopheryl acetate) -0.01 - 0.01 °
Acetic acid (HS-GC) ≤ 5000 ppm
Heptane (HS-GC) ≤ 5000 ppm
Methanol (HS-GC) ≤ 3000 ppm
Pyridine (HS-GC) ≤ 200 ppm
Toluene (HS-GC) ≤ 890 ppm
Other residual solvents (Ph.Eur./USP/ICH) excluded by production process
Loss on drying (130 °C, vacuum, 4 h) ≤ 5.0 %
Total aerobic microbial count (TAMC) ≤ 100 CFU/g
Total combined yeasts/moulds count (TYMC) ≤ 100 CFU/g
Bile-tolerant gram-negative bacteria (absent in 1 g) passes test
Candida albicans (absent in 1 g) passes test
Escherichia coli (absent in 1 g) passes test
Pseudomonas aeruginosa (absent in 1 g) passes test
Salmonella (absent in 10 g) passes test
Staphylococcus aureus (absent in 1 g) passes test
corresponds to Ph Eur, BP, USP

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR