DL-α-Tocopherol (Vitamin E), dry powder sprayed, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS

5.00878.9024
5.00878.9024EA 0 EUR
5.00878.9024
DL-α-Tocopherol (Vitamin E), dry powder sprayed, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
DL-α-Tocopherol

Vitamin E (50 g all-rac-α-tocopheryl acetate in 100 g substance)

Formula: C₂₉H₅₀O₂
MDL Number: MFCD00072051
CAS Number: 10191-41-0
EINECS: 233-466-0

Order Now

Specification Test Results

all-rac-α-Tocopheryl acetate content (GC, Ph Eur) 50.0 - 57.5 %
Identity (GC) passes test
As (Arsenic) ≤ 0.0003 %
Cu (Copper) ≤ 50 ppm
Pb (Lead) ≤ 10 ppm
Zn (Zinc) ≤ 25 ppm
Ethanol (HS-GC) ≤ 5000 ppm
Acetic acid (HS-GC) ≤ 5000 ppm
Heptane (HS-GC) ≤ 5000 ppm
Methanol (HS-GC) ≤ 3000 ppm
Pyridine (HS-GC) ≤ 200 ppm
Toluene (HS-GC) ≤ 890 ppm
Residual solvents (ICH Q3C) excluded by production process
Loss on drying ≤ 5.0 %
Total aerobic microbial count (TAMC) ≤ 1000 CFU/g
Total combined yeasts/moulds count (TYMC) ≤ 100 CFU/g
Bile-tolerant gram-negative bacteria (absent in 1 g) passes test
Candida albicans (absent in 1 g) passes test
Escherichia coli (absent in 1 g) passes test
Pseudomonas aeruginosa (absent in 1 g) passes test
Salmonella (absent in 10 g) passes test
Staphylococcus aureus (absent in 1 g) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR