Orthophosphoric acid 85,0, LiChropur™ HPLC, Supelco®

Supplier: Merck
Danger

Synonyms: Phosphoric acid

MQ100

5.43828.0250 5.43828.0100
5.43828.0250EA 249 EUR
5.43828.0250 5.43828.0100
Orthophosphoric acid 85,0, LiChropur™ HPLC, Supelco®
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 158
Melting Pt: 21
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 1805
ADR: 8 III
Merck Index: 14,07344

Order Now

Specification Test Results

Assay (Alkalimetric) ≥85,0%
Identity Passes Test
Colour ≤10 Hazen
Insoluble matter ≤10 ppm
Aluminium (Al) ≤0,020 ppm
Calcium (Ca) ≤0,050 ppm
Copper (Cu) ≤0,025 ppm
Iron (Fe) ≤0,020 ppm
Pottasium (K) ≤0,050 ppm
Magnesium (Mg) ≤0,020 ppm
Sodium (Na) ≤0,10 ppm
HPLC gradient suitability test (220, 254 and 280 nm) ≤10 ppm
Chromophore purity ≥99,999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR