Multiple adapters
Supplier: Lenz Laborglas GmbH & CO.KG
05000222 05000414 05000444 05000111
5000222.EA 46.8 EUR
5000222. 5000414. 5000444. 5000111.
Multiple adapters
Distillation Components Multiple Distillation Adapters
DURAN®, borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two standard ground 29/32 sockets and one standard ground 29/32 cone. Side neck inclined upwards at an angle of 105°.
Order Now
SPECIFICATIONS
Environmentally Preferable