Multiple adapters
Supplier: Lenz Laborglas GmbH & CO.KG
05015011 05015024 05015044 05015023 05015033
5015011.EA 92 EUR
5015011. 5015024. 5015044. 5015023. 5015033.
Multiple adapters
Distillation Components Multiple Distillation Adapters
DURAN® Borosilicate glass 3.3
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With 3 NS sockets and 1 NS cone, with two parallel necks and one angled neck (45°).
Order Now
SPECIFICATIONS
Environmentally Preferable