Organic reference standard, Senecionine, analytical standard, Supelco®

Supplier: Merck

50351-5MG
50351-5MGEA 439 EUR
50351-5MG
Organic reference standard, Senecionine, analytical standard, Supelco®
Standards Organic Standards

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c504e9941a83b5fcd3ade2ade5ed21d096078c5&vid=6c504e9941a83b5fcd3ade2ade5ed21d096078c5&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Description Senecionine
Environmentally Preferable
CAS No. 130-01-8
Formula C₁₈H₂₅NO₅
Storage temperature 2 - 8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR