Benzyl acetate, Supelco®

Supplier: Merck


MQ100

50475-5ML
50475-5MLEA 83.3 EUR
50475-5ML
Benzyl acetate, Supelco®
Benzyl acetate
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 206
Melting Pt: –51 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 95
Storage Temperature: Ambient
MDL Number: MFCD00008712
CAS Number: 140-11-4
EINECS: 205-399-7
Merck Index: 13,01125
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=747191ff73f5e7c03e03f4fd8eda29ef4d0415a8&vid=92dde70d8e3ad3be758ba902166fb82c36c8f0f2&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR