Distilling bridges with Liebig condenser with glass olive or GL thread connection
Supplier: Lenz Laborglas GmbH & CO.KG
05050304 05050404 05050301 05050201 05050211 05050311 05050314
5050304.EA 152 EUR
5050304. 5050404. LENZ05050301 5050201. LENZ05050211 LENZ05050311 LENZ05050314
Distilling bridges with Liebig condenser with glass olive or GL thread connection
Distillation Components Distillation Bridges
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling links with Liebig condenser, outlet with NS cone, water connection, with socket NS 14/23 for distillation thermometers of 55 mm built-in length.
Order Now
SPECIFICATIONS
Environmentally Preferable