Distilling bridges, Claisen
Supplier: Lenz Laborglas GmbH & CO.KG
05057201 05057301 05057304 05057404
5057201.EA 181 EUR
5057201. 5057301. 5057304. 5057404.
Distilling bridges, Claisen
Distillation Components Distillation Bridges
DURAN®, borosilicate 3.3 glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Still heads with Liebig condenser and Claisen attachment. With NS 14/23 socket for distillation thermometer, with 55 mm total length and glass vacuum tube connector. Connection for coolant water via tube connector.