Supelco® TraceCERT® Organic Reference Standard, ʟ-Tryptophan
Supplier: Merck
51145-100MG
51145-100MGEA 88.5 EUR
51145-100MG
Supelco® TraceCERT® Organic Reference Standard, ʟ-Tryptophan
Standards Organic Standards
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7689a1471ec8fe46180651f9a7c59230b514992d&vid=7689a1471ec8fe46180651f9a7c59230b514992d&prid=web&clid=SAPDAM