Grade 114 qualitative filter papers, Whatman™
Supplier: Whatman products (Cytiva)
1114-090 1114-125 1114-150 1114-185 1114-240 1114-400 1114-930
512-0620EA 16.4 EUR
512-0620 512-0621 1114150. 512-0622 512-0623 512-0624 WHAT1114-930
Grade 114 qualitative filter papers, Whatman™
Filters Filter Papers Qualitative Filter Paper
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin.
  • Only half the thickness of Grade 113 and suitable for coarse or gelatinous precipitates
  • Smooth surface for easy recovery of precipitates
Normal qualitative applications will not introduce any significant impurities into the filtrate. The resins do, however, contain nitrogen so these grades should not be used in Kjeldahl estimations, etc. Some wet strengthened grades are available in folded (prepleated) forms.