Filtrate centrifugal ultrafilter, Vivaspin®
Supplier: Sartorius
13239-E 13229-E 13249-E 13279-E 13269-GE
512-3532EA 118 EUR
512-3532 512-3531 512-3533 512-3535 SART13269-GE
Filtrate centrifugal ultrafilter, Vivaspin®
Filters Centrifugal Filters
These centrifugal devices are ideal for the concentration or ultrafiltration of difficult to filter samples, even whole blood or cell lysates.
  • PS body
  • Styrene acrylonitrile (SAN) filtrate vessel
  • PE concentrator cap
  • Filtrate is a ready-to-use ultrafiltration unit for up to 2,5 ml samples
  • Unhindered passage of low molecular weight solutes prior to ultrafiltrate analysis
  • Rapid protein removal from biological samples
  • Recover samples directly, without the need to remove any caps or disassemble the unit
They may be used for removal of low molecular weightMore Product Information
Order Now
SPECIFICATIONS
Filtration area 0,79 cm²
Environmentally Preferable
Max. RCF - swing-out rotor 2500×g
Max. RCF - fixed angle rotor 2000×g
Ø×H 14×93 mm
Sample volume 500 - 2500 µl
Dead stop volume 100 µl
Hold-up volume <5 µl