Filter plates, 96-well, for solvent filtration, AcroPrep™ Advance
Supplier: Cytiva (Formerly Pall Lab)
8047 8048 8247 8148 8147 8248 8582 8584 8682 8684 8782 8784 8586 8588 8686 8688
738-0109EA 418 EUR
738-0109 738-0110 738-0131 738-0122 738-0121 738-0132 513-0163 513-0164 514-1096 514-1097 514-1098 514-1099 513-0166 513-0168 513-0167 513-0169
Filter plates, 96-well, for solvent filtration, AcroPrep™ Advance
Microplates Filter Plates
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes that provide the best possible performance for a wide range of sample preparation and high throughput screening applications. Available with a choice of PTFE or PTFE membrane.
  • Chemically resistant materials provide a stable platform to process samples in organic solvents
  • Low, non specific binding: inert construction materials ensure complete sample recovery
  • Well geometry results in faster, more uniform filtration rates across the plate with reduced hold-up
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable