Sampling head, MD8 Airscan®
Supplier: Sartorius
16746-01-SHB
514-1293EA 7480 EUR
514-1293
Sampling head, MD8 Airscan®
Filters Bottle Top Filters
Sampling head with bayonet connection.
  • Flexible operating conditions
  • Generates a vacuum and pumps a defined air sample volume from the ambient air
The bayonet connector accommodates the filter adapter. Sampling heads with a bayonet connection are particularly suitable for applications where the sampling head is not installed in a system.
Ordering information: Supplied without accessories.