HEPA capsules
Supplier: Cytiva (Formerly Pall Lab)
12144
514-4092EA 253 EUR
514-4092
HEPA capsules
Filters Venting Filters
Designed to provide bacteria-free air (HEPA: High Efficiency Particulate free Air) for sterile applications. Porous membrane delivering high airflow rates at low differential pressure and extends filter life.
  • Can be used for non sterilising liquid prefiltration
Versapor® membrane, PP housing.
Ensures particulate free air with 99,97% retention of 0,3 µm DOP aerosol (ASTM D 2986-71).