Grade 2589 C filter papers for technical use, Whatman™
Supplier: Whatman products (Cytiva)
10343876
515-8080EA 76.1 EUR
515-8080
Grade 2589 C filter papers for technical use, Whatman™
Filters Filter Papers Qualitative Filter Paper
Thick filter with medium to slow filtration speed, high wet strength, and good retention for smaller particles.
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application. The applications range from the filtration of beverages to the purification of electroplating baths.