Filter funnels, Sentino®
Supplier: Cytiva (Formerly Pall Lab)
4870 4871
516-0666EA 112 EUR
516-0666 516-0667
Filter funnels, Sentino®
Funnels Membrane Filter Funnels
The Sentino® filter funnel and adapter are designed with a press-fit seal, membrane guides and forceps access point allowing for simple set-up and membrane removal. This will speed up the process time in the laboratory.
  • Easy placement
  • Risk reduction
  • Regulatory compliance
  • Allows easy and accurate sample measurement
  • Available in 100 and 250 ml capacity to accommodate a variety of sample sizes
  • Inert polypropylene construction to preserve the integrity of the sample
  • Base adapts for use with Sentino® microbiology pump or standard manifold
Easy to use,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable