Formaldehyde 10% in aqueous solution stabilised

Supplier: VWR Chemicals
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

Stabilisation: 0,5-1,5 % Stabilised with methanol

5167.1000EA 51.8 EUR
5167.1000
Formaldehyde 10% in aqueous solution stabilised
Formaldehyde
Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: -19.3
Melting Pt: -92
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 3334
ADR: 9 null
Merck Index: 13,04259

Order Now

Specification Test Results

Assay (Formaldehyde) 8 - 12 %
pH (20°C) 8.2 - 8.4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR