Centrifuges, benchtop, ventilated/refrigerated, ROTINA 380 / 380 R
Supplier: Hettich
1701 1706
521-1153EA 5630 EUR
521-1153 521-1154
Centrifuges, benchtop, ventilated/refrigerated, ROTINA 380 / 380 R
Centrifuges Benchtop Centrifuges
The high performance benchtop centrifuges ROTINA 380 and ROTINA 380 R impress with state of the art technology, large capacity, extremely easy handling and exceptionally smooth and quiet running.
  • Smooth and quiet running, brushless, maintenance-free induction drive
  • Housing and lid made of metal for speed stability, centrifuging chamber and inner lid cover made of stainless steel
  • Ergonomically arranged controls and displays, easy to operate
  • Special lid locking device enables effortless closing and opening of the lid with one hand
  • Safety features:
More Product Information