Centrifuges, compact, EBA 200 / 200 S
Supplier: Hettich
1802 1800
521-1930EA 2780 EUR
521-1930 521-1928
Centrifuges, compact, EBA 200 / 200 S
Centrifuges Benchtop Centrifuges
These compact centrifuges come with a fixed angle rotor and are ideal for low numbers of samples.
  • Both models are equipped with 8-place angle rotor which holds tubes up to 15 ml
  • Maintenance-free frequency drive
  • Compact size saves space on the benchtop
EBA 200 is ideal for medical diagnostics in clinical environments. The high speed of the EBA 200 S enables shorter centrifugation time so that plasma with a low platelet count can be obtained for analysis in just a few minutes.
Ordering information: Both centrifuges are supplied with fixed-angleMore Product Information