Bioprocessing centrifuges, refrigerated, floor standing, Thermo Scientific™ Sorvall™ BIOS 16 and Sorvall™ BIOS A

Supplier: SORVALL
75007685 75007697 75007698 75007699
521-2685EA 0 EUR
521-2685 521-2686 521-2687 521-2688
Bioprocessing centrifuges, refrigerated, floor standing, Thermo Scientific™ Sorvall™ BIOS 16 and Sorvall™ BIOS A
Centrifuges Floor-Standing Centrifuges
These large capacity centrifuges are designed for high throughput bioprocessing and support a broad array of applications: Biologics, vaccines, biofuels, veterinary medicines, recombinant proteins and raw materials.

  • Glove-friendly interface with bright and highly visible display simplifies run set-up
  • Automatic 'front to back' centrifuge door opening and closing with the auto-door function
  • Auto-ID instant rotor identification: Immediate identification of a rotor when secured in the centrifuge chamber with rotor specifications automatically loaded into the
More Product Information
Order Now

SPECIFICATIONS

Model Sorvall™ BIOS 16 Sorvall™ BIOS 16 Sorvall™ BIOS A Sorvall™ BIOS A
Environmentally Preferable
Max. capacity 8×2000 ml 8×2000 ml 10×1000 ml 10×1000 ml
Max. RCF 8500 ×g 8500 ×g 12000 ×g 12000 ×g
Max. speed 5400 min⁻¹ 5400 min⁻¹ 6250 min⁻¹ 6250 min⁻¹
Acceleration/braking profiles 11/12 11/12 11/12 11/12
Control Touch screen interface Touch screen interface Touch screen interface Touch screen interface
Programs 99 99 99 99
Sound level <62 dB(A) <62 dB(A) <62 dB(A) <62 dB(A)
Timer 99 h, hold 99 h, hold 99 h, hold 99 h, hold
Temperature –20...+40 °C –20...+40 °C –20...+30 °C –20...+30 °C
Motor DuraFlex, high torque, brushless DuraFlex, high torque, brushless DuraFlex, high torque, brushless DuraFlex, high torque, brushless
Power supply 200 - 240 V, 50/60 Hz, single phase 380 - 415 V, 50 Hz, 3 phase 200 - 240 V, 50/60 Hz, single phase 380 - 415 V, 50 Hz, 3 phase
W×D×H 816×990×1015 mm 816×990×1015 mm 816×990×1015 mm 816×990×1015 mm
Weight 475 kg 495 kg 481 kg 466 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR