Megafuge™ ST4/ST4R Plus centrifuges (IVD-marked)
Supplier: Thermo Scientific
75016079 75009503 75009518 75016060 75009519
521-2740EA 19210 EUR
521-2740 521-2725 521-2727 521-2734 HERA75009519
Megafuge™ ST4/ST4R Plus centrifuges (IVD-marked)
Centrifuges Benchtop Centrifuges
Ideal for routine processing, these centrifuges combine exceptional capacity, ergonomics, and value with market-leading capabilities including unparalleled productivity and throughput, as well as exceptional ergonomics.
  • Patented lid locking technology
  • Easily adjustable speed, time, and temperature during the run
  • Modern, intuitive touchscreen interface enables easy access and programming for faster results
  • Lighthouse mode: Easily monitor run progress from across the laboratory with enlarged time and tracker bar, end-of-run alerts can be programmed with
More Product Information