H-FLEX HS4 windshield swinging bucket rotor and accessories
Supplier: Thermo Scientific
75003330 75003338 75003339
521-3440EA 4560 EUR
521-3440 521-3441 521-3442
H-FLEX HS4 windshield swinging bucket rotor and accessories
Centrifuges Floor-Standing Centrifuges
Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor at 7197xg in 4 L High Performance General Purpose Centrifuges.
  • SMARTSpin™ technology
  • Space saving design
Swing bucket rotor enables fast Auto-Lock, one-handed rotor exchanges. Spins blood tubes, conical tubes and microplates without changing rotor or buckets. Results are faster without the need to change hardware. Limit rotor maintenance and contamination potential with slide-coat technology eliminatingMore Product Information