Cryofuge™ 8/16 Blood Banking Centrifuges with GreenCool Technology
Supplier: Thermo Scientific
75008600 75008603 75008610 75008615 75008618
521-4482EA 53270 EUR
521-4482 521-4483 521-4484 521-4485 521-4486
Cryofuge™ 8/16 Blood Banking Centrifuges with GreenCool Technology
Centrifuges Floor-Standing Centrifuges
Thermo Scientific™ Cryofuge™ 8/16 Blood Banking Centrifuges with GreenCool Technology bring enhanced sustainability to blood processing centres. Featuring a natural refrigerant (CO2) cooling system, they offer reduced energy consumption and heat output, have short cool-down times and maintain excellent temperature stability for precise results whilst complying with global standards.
  • Global Warming Potential (GWP) of 1, up to 14% lower energy consumption
  • Large capacity of up to 16×550 ml blood bags (Cryofuge 16)
  • Auto-ID instant rotor identification
More Product Information