Eppendorf Tubes® Microtubes with Screw Cap, 5,0 ml
Supplier: EPPENDORF
0030122.313 0030122.321 0030122.305 0030122.402 0030122.330 0030122.364 0030113.500 0030122.518
525-0981EA 67.5 EUR
525-0981 525-0982 525-0980 EPPE0030122.402 EPPE0030122.330 EPPE0030122.364 EPPE0030113.500 525-1615
Eppendorf Tubes® Microtubes with Screw Cap, 5,0 ml
Tubes Microcentrifuge Tubes
Eppendorf Tubes® with screw cap are an ideal choice when working with medium-sized sample volumes (0,5 to 5,0 ml). SafeCode variants enhance sample identification, while the BioBased option, manufactured using renewable resources, provides an eco-friendly alternative without compromising performance.
  • Polypropylene (PP) tubes, high-density polyethylene (HDPE) screw caps
  • Ensures leak-free storage and protects sample integrity
  • Suited for a wide range of laboratory tasks, enhancing flexibility
  • High-quality materials for long-lasting, robust performance
  • Clear,
More Product Information
Order Now
SPECIFICATIONS
Max. RCF 25000×g 25000×g 25000×g 25000×g
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact,Sustainable Materials
Operating temperature range −86...+100 °C −86...+100 °C −86...+100 °C −86...+100 °C
Volume 5,0 ml 5,0 ml 5,0 ml 5,0 ml
Environmental certification Renewable Energy Certificate, ISO 14001 Renewable Energy Certificate, ISO 14001. ISCC PLUS, Renewable Energy Certificate, ISO 14001, ACT Label