Screw caps for microcentrifuge tubes
Supplier: Thermo Fisher Scientific
3471 3471B 3471G 3471O 3471R 3471Y 525 3471T 3471TA 3471TAS 3471TB 3471TBLK 3471TBLKS 3471TBS 3471TG 3471TGS 3471TMIXS 3471TO 3471TOS 3471TPS 3471TR 3471TRS 3471TS 3471TV 3471TVS 3471TWS 3471TY 3471TYS 3471AS 3471BLK 3471BLKS 3471BS 3471GS 3471MIX 3471MIXS 3471OS 3471P 3471PS 3471RS 3471S 3471V 3471VS 3471W 3471WS 3471YS
MLBP3471EA 83 EUR
MLBP3471 MLBP3471B MLBP3471G MLBP3471O MLBP3471R MLBP3471Y MLBP525 525-1385 525-1386 525-1387 525-1388 525-1389 525-1390 525-1391 525-1392 525-1393 525-1395 525-1396 525-1397 525-1399 525-1400 525-1401 525-1407 525-1408 525-1409 525-1415 525-1416 525-1417 525-1373 525-1374 525-1375 525-1376 525-1377 525-1378 525-1379 525-1380 525-1381 525-1382 525-1383 525-1384 525-1418 525-1419 525-1420 525-1421 525-1422
Screw caps for microcentrifuge tubes
Closures Test/Sample Tube Closures
Thermo Scientific™ screw cap microtube caps with O-ring are designed to create a secure seal with the Thermo Scientific™ microtube for sample preparation, transfer and storage applications. Tethered and non-tethered cap versions are available in a rainbow of colours.
  • Non sterile: Certified DNA, Rnase/Dnase, Pyrogen and Endotoxin free
  • E-beam sterile: Certified DNA, Rnase/Dnase, Pyrogen, Endotoxin, PCR inhibitor, and Bioburden free