Centrifuge tubes closure, Nalgene®
Supplier: Thermo Fisher Scientific
DS3111-0016 DS3111-0017 DS3111-0018 DS3111-0025 DS3111-0029 DS3111-0030 DS3111-0032
525-2932EA 77.6 EUR
525-2932 525-2933 525-2934 525-2936 525-2937 525-2938 525-2939
Centrifuge tubes closure, Nalgene®
Closures Test/Sample Tube Closures
PP, autoclavable.
  • Designed for both Nalgene® round bottom and conical centrifuge tubes
  • A tab facilitates the handling of the closure
Ordering information: 525-2937 is for 50 ml tubes with rim and round bottom, 525-2938 is for 50 ml round bottom, rimless tubes.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement