Centrifuge tubes, conical
Supplier: DELTALAB
429910 429920 429900 429901 429926 429927 429946 429927.10 429927.25
525-4214EA 155 EUR
525-4214 525-4215 525-4216 525-4217 525-4218 525-4219 DELT429946 DELT429927.10 DELT429927.25
Centrifuge tubes, conical
Tubes Centrifuge Tubes
Tubes made of PP, translucent, with blue HDPE screw cap.
  • Continuous thread
  • External moulded graduations in 0,5 ml increments
  • Excellent temperature resistance
  • Excellent chemical resistance
Suitable for centrifugation tests in immunology, microbiology, more.