Distilling receivers, Bernauer
Supplier: Lenz Laborglas GmbH & CO.KG
05288101 05288001 05288002 05288004 05288102 05288104
5288101.EA 136 EUR
5288101. 5288001. 5288002. 5288004. 5288102. 5288104.
Distilling receivers, Bernauer
Distillation Components Distillation Receivers
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling receivers acc. to Bernauer (modified), with three cones NS 14/23, vacuum connection with glass olive. With flasks 100 ml capacity or without flasks available.
Order Now
SPECIFICATIONS
Environmentally Preferable