Potassium dihydrogen phosphate, Millipore®

Supplier: Merck Millipore Calbiochem

529568-1
529568-1EA 318 EUR
529568-1
Potassium dihydrogen phosphate, Millipore®
Potassium dihydrogen phosphate
Formula: KH₂PO₄
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 253 °C
Density: 2,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR