Supelco® Ascentis® Express RP-Amide HPLC and UHPLC Columns
Supplier: Merck
53910-U 53911-U 53912-U 53913-U 53914-U 53915-U 53916-U 53917-U 53918-U 53919-U 53921-U 53922-U 53923-U 53929-U 53931-U 53514-U 53516-U 53519-U 53797-U 50732-U 50733-U 50734-U 50735-U 50737-U 50739-U 50747-U 50749-U 50751-U 50752-U 50753-U 50758-U 50759-U 50767-U 50768-U 50769-U 50773-U 50774-U 50775-U 50776-U 50777-U 50779-U 51567-U 51568-U 51569-U 51571-U 51576-U 51577-U 51582-U 51583-U 51588-U 51589-U 51594-U 51595-U
53910-UEA 491 EUR
53910-U 53911-U 53912-U 53913-U 53914-U 53915-U 53916-U 53917-U 53918-U 53919-U 53921-U 53922-U 53923-U 53929-U 53931-U 53514-U 53516-U 53519-U 53797-U 50732-U 50733-U 50734-U 50735-U 50737-U 50739-U 50747-U 50749-U 50751-U 50752-U 50753-U 50758-U 50759-U 50767-U 50768-U 50769-U 50773-U 50774-U 50775-U 50776-U 50777-U 50779-U 51567-U 51568-U 51569-U 51571-U 51576-U 51577-U 51582-U 51583-U 51588-U 51589-U 51594-U 51595-U
Supelco® Ascentis® Express RP-Amide HPLC and UHPLC Columns
Chromatography Columns
Ascentis Express RP-Amide high-speed, high-performance liquid chromatography columns are based on Fused-Core® particle design. The fused core partical provides a thin porous shell of high-purity silica surrounding a solid silica core.
This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0,4 μm thick porous shell. The one-step bonded, end-capped, amide-based polar-embedded stationary phase of Ascentis Express RP-Amide provides, a stable, reversed phase packing with decreased hydrophobic character.
Ascentis Express RP-Amide can be used forMore Product Information