Extractors for specific light solvents
Supplier: Lenz Laborglas GmbH & CO.KG
05432037 05432049 05432058
5432037.EA 721 EUR
5432037. 5432049. 5432058.
Extractors for specific light solvents
Extractors Liquid-Liquid Extractors
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of liquids with specific light solvents.
Order Now
SPECIFICATIONS
Environmentally Preferable