Extractors for specific heavy solvents
Supplier: Lenz Laborglas GmbH & CO.KG
05433037 05433049 05433058
5433037.EA 619 EUR
5433037. 5433049. 5433058.
Extractors for specific heavy solvents
Extractors Liquid-Liquid Extractors
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of liquids with specific heavy solvents.
Order Now
SPECIFICATIONS
Environmentally Preferable