Septa 20 mm for crimp caps, Chromacol™
Supplier: CHROMACOL
20-LLX
548-0743EA 129 EUR
548-0743
Septa 20 mm for crimp caps, Chromacol™
Septa
High purity 20 mm silicone/PTFE seal for liquid-liquid extraction and trace analysis. PTFE thickness 0,02 mm and silicone 3 mm.