Gas-tight syringes, 10 to 500 µl
Supplier: Trajan Scientific and Medical
002200 002208 002250 002252 002455 002500 003200 003250 004200 004229 004230 004250 004279 005200 005229 005230 005250 005279 006200 006229 006230 006250 006279 007200 007229 007230 007250 002202 004232 005232 006232 007232 007250CAL 002200CAL 003200CAL 004200CAL 006200CAL 005200CAL 007200CAL
002200.EA 74.8 EUR
002200. 549-0445 002250. 562-1031 549-0448 002500. 003200. 003250. 004200. 549-0301 549-0302 004250. 549-0304 005200. 549-0311 548-0940 005250. 549-0313 006200. 549-0319 549-0320 006250. 549-0321 007200. 548-0945 548-0946 007250. 548-0935 548-0938 548-0941 548-0942 548-0947 SGEA007250CAL SGEA002200CAL SGEA003200CAL SGEA004200CAL SGEA006200CAL SGEA005200CAL SGEA007200CAL
Gas-tight syringes, 10 to 500 µl
Syringes Syringes for Chromatography
Constructed from borosilicate glass, PTFE plunger tips and Kel-F® or PTFE Luer cones where applicable.
  • Assembled with an interference fit between plunger tip and barrel to effect the gas sealing properties of the syringe
  • Plungers are replaceable on these syringes (plungers on request)
Barrel OD: 10 µl = 6,5 mm; 25 to 500 µl = 8 mm