VWR®, Short Thread Vials ND9
Supplier: VWR Collection
548-0030AEA 346 EUR
548-0030A 548-9011A 548-9012A 548-2495A 548-1419A 548-3343A 548-2497A 548-2498A 548-0177A 548-2496A 548-1848A 548-3275A 548-1092A 548-3309A 548-1055A 548-3276A 548-0081A 548-1373A 548-2095A 548-1853A 548-0910A 548-0028A 548-0029A 548-1442A 548-3201A 548-8011A 548-8012A 548-8013A 548-2533A 548-1422A 548-3342A 548-1396A 548-0854A 548-0907A 548-1421A 548-1056A 548-0706A 548-2488A 548-0911A 548-3255A 548-3213A 548-0912A 548-3253A 548-2489A 548-2491A 548-0426A 548-0909A 548-0427A 548-3352A 548-0908A 548-2490A 548-3259A 548-2002A 548-2125A 548-3047A 548-2364A 548-1363A 548-1457A 548-1083A 548-1471A 548-2144A 548-2121A 548-2094A 548-0428A 548-0894A 548-0440A 548-0452A 548-0455A 548-0453A 548-0450A 548-2098A 548-0855A 548-1417A 548-1488A 548-1851A 548-1931A 548-1955A 548-2134A 548-2172A 548-2359A 548-2372A 548-2420A 548-2421A 548-2482A 548-2593A
VWR®, Short Thread Vials ND9
Vials
Short thread ND9 vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polypropylene seal styles and septa materials.
  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
  • Easy filling (wide opening)
Short thread ND9 vials constructed from type I borosilicate glass with low metal content. Vials are available in amber or clear version with a flat bottom to ensure compatibility with glass and plastic inserts. Associated screw caps mostly available with assembled septa areMore Product Information