VWR® PFAS Short Thread or Snap Ring Caps
Supplier: VWR Collection
548-7010AEA 22.5 EUR
548-7010A 548-7006A 548-7003A 548-7012A
VWR® PFAS Short Thread or Snap Ring Caps
Closures Chromatography Vial Closures
PFAS PP short thread caps or PE snap ring caps, light green, with hole. White silicone/transparent polypropylene, for PFAS analysis.
  • PFAS free caps for the PFAS sample process
  • PFAS can be found in:
  • Environmental monitoring
  • Food and water quality testing
  • Product testing
  • Industrial quality assurance
  • Commercial household cleaning products
  • Food packaging
PFAS stands for per- and polyfluoroalkyl substances, and are colloquially known as ‘forever chemicals’. They are a group of man-made chemicals that have been widely used in various industrial and consumer
More Product Information