TLC plate heater III
Supplier: CAMAG
022.3306UK
552-0351EA 3190 EUR
552-0351
TLC plate heater III
Heaters Hotplates
The TLC plate heater III is designed for heating TLC plates to a given temperature, while ensuring homogenous heating across the plate. TLC plate heater III has a CERAN® heating surface which is resistant to all common reagents and is easily cleaned. The 200×200 mm heating surface has a grid to facilitate correct positioning of the TLC plate.
  • Actual temperature is displayed digitally
  • Temperature is selectable between 25 and 200 °C
  • Plate heater is protected from overheating
Order Now
SPECIFICATIONS
Environmentally Preferable