Size exclusion chromatography columns, OHpak SB series
Supplier: Resonac
F6429105 F6709430 F6429102 F6429101 F6429103 F6429104 F6429100 F6429106 F6429108 F6709431
554-1747EA 1809 EUR
554-1747 554-1754 554-1750 554-1751 554-1749 554-1748 554-1752 554-1746 554-1745 554-1753
Size exclusion chromatography columns, OHpak SB series
Chromatography Columns
SB-800 HQ are polyhydroxymethacrylate-based SEC columns for molecular weight distribution analysis of water-soluble polymers. The solvent can be substituted with DMF (except for SB-802 HQ and SB-807 HQ columns), enabling the analysis of polar polymers.
  • Polymer-based SEC (GFC) columns for aqueous systems
  • Support a wide range of molecular weight
  • SB-807 HQ with an exclusion limit of 500000000 is available for the analysis of macromolecules
* Estimated value