HPLC columns, Accucore™ Urea-HILIC
Supplier: Thermo Fisher Scientific
27726-102130
554-2329EA 1260 EUR
554-2329
HPLC columns, Accucore™ Urea-HILIC
Chromatography Columns
These columns offer a unique HILIC selectivity and low ion exchange activity. Based on Core Enhanced Technology, they provide fast, high resolution separations without the elevated back pressures required by sub-2 µm particles.
  • Fast separations with high resolution
  • Low back pressure
  • Bonded hydrophilic stationary phase
Rugged 2,6 µm solid core particles enable fast, high-resolution separations at low backpressures. The Urea-HILIC bonded hydrophilic stationary phase offers unique selectivity and has lower ion exchange activity compared to other HILICMore Product Information