Ion chromatography columns, anion analysis
Supplier: Resonac
F6995240 F6700400 F6709616 F6995243 F6709620 F6995244 F6995245 F6995260 F6709626 F6709625
554-2906EA 1444 EUR
554-2906 554-2880 554-2897 554-2907 554-2898 554-2908 554-2909 554-2911 554-2901 554-2900
Ion chromatography columns, anion analysis
Chromatography Columns
NI-424, I-524A
- Columns for anion analysis with non suppressor method
- NI-424 supports simultaneous analysis of fluoride and phosphate ions
SI-90, SI-50
- Columns for anion analysis with suppressor method
- Suitable for the quantitative analysis of fluoride ion
- SI-50 separates target inorganic anions from organic acids
- Not interfered by the system peak derived from carbonate
SI-52
- Columns for the analysis of oxyhalides
- Supports simultaneous analysis of oxyhalides and general inorganic ions