Capillary columns, TraceGOLD™ TG-200MS
Supplier: Thermo Fisher Scientific
26084-1420 26084-1430 26084-2240 26084-2960 26084-2970
555-8896EA 1020 EUR
555-8896 555-8895 555-8893 555-8890 555-8889
Capillary columns, TraceGOLD™ TG-200MS
Chromatography Columns
TraceGOLD™ TG-200MS GC columns are exceptionally inert mid-polarity columns with selectivity and elution order optimised for difficult separations.
  • Polar phase, Trifluoropropylmethyl polysiloxane solid phase resolves compounds that phenyl and cyano phases cannot
  • Outstanding thermal stability and low bleed
  • Suitable for use with sensitive detectors including ECDs, NPDs and MS
  • Equivalent to USP G6 phase
  • Confirmation column in combination with another GC column